Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Hexyl Alcohol
SCI_ED

Catalog No. S25701A Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S25701A 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S25701A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 111-27-3
Density 0.814g/mL
Assay Percent Range >95% w/v
Packaging Solution
Molecular Formula C6H14O
Linear Formula CH3(CH2)4CH2OH
Quantity 500 mL
Synonym 1-hexanol, hexyl alcohol, hexanol, n-hexanol, n-hexyl alcohol, 1-hexyl alcohol, 1-hydroxyhexane, amylcarbinol, caproyl alcohol, pentylcarbinol
InChI Key ZSIAUFGUXNUGDI-UHFFFAOYSA-N
SMILES CCCCCCO
IUPAC Name hexan-1-ol
Molecular Weight (g/mol) 102.177
PubChem CID 8103
ChEBI CHEBI:87393
Formula Weight 102.18
Chemical Name or Material Hexyl Alcohol
Show More Show Less
Safety handling Check Alert storage code Red
ShelfLife 5 years
DOTInformation DOT Class 3, III, : Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.