Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexyl Alcohol Pract., 96%, Thermo Scientific™

Catalog No. AC411655000
Change view
Click to view available options
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC411655000 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC411655000 Supplier Thermo Scientific Chemicals Supplier No. 411655000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 111-27-3
Density 0.8136g/mL
Infrared Spectrum Authentic
Assay Percent Range 96%
Molecular Formula C6H14O
Refractive Index 1.4170 to 1.4190 (20°C, 589nm)
Linear Formula CH3(CH2)5OH
Quantity 500g
MDL Number MFCD00002982
Beilstein 01, 407
Merck Index 15, 4734
Synonym 1-hexanol, hexyl alcohol, hexanol, n-hexanol, n-hexyl alcohol, 1-hexyl alcohol, 1-hydroxyhexane, amylcarbinol, caproyl alcohol, pentylcarbinol
InChI Key ZSIAUFGUXNUGDI-UHFFFAOYSA-N
SMILES CCCCCCO
IUPAC Name hexan-1-ol
Molecular Weight (g/mol) 102.177
PubChem CID 8103
ChEBI CHEBI:87393
Formula Weight 102.18
Percent Purity ≥95%
Chemical Name or Material Hexyl alcohol, pract., 96%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Harmful if swallowed. Harmful in contact with skin. Causes serious eye irritation. Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Call a POISON CENTER or doctor/physician if you feel unwell. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
EINECSNumber 203-852-3
RTECSNumber MQ4025000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.