Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexyl Hexanoate, Analytical Standard, ≥98.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111015012 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5012 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5012 Supplier MilliporeSigma Supelco Supplier No. 182821ML

missing translation for 'validMainframeMsg'

Specifications

CAS 6378-65-0
Density 0.863 g/mL (at 25°C)
Physical Form Neat
Linear Formula CH3(CH2)4 CO2(CH2)5 CH3
MDL Number MFCD00053808
Melting Point -55°C
Boiling Point 245°C to 246°C
Synonym Hexyl caproate
Quantity 1 mL
Formula Weight 200.32
Percent Purity ≥98.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.