Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Hexyl isocyanate

Catalog No. 4148835ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-488-35ML 5 mL
41-488-325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 41-488-35ML Supplier Sigma Aldrich Supplier No. 4148835ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Hexyl isocyanate has been investigated for its antigenicity in the guinea pig animal model of hapten-specific respiratory hypersensitivity. polymerization of n-hexyl isocyanate using rare earth tris(2,6-di-tert-butyl-4-methylphenolate) [ln(oar)3] as initiator has been studied.hexyl isocyanate may be employed for the synthesis of following:
  • Rigid-rod, helical isocyanate-based macromonomers
  • Rod-coil-rod triblock copolymers with 2-vinylpyridine, by living anionic polymerization
  • Chiral poly(n-hexyl isocyanate) (phic) macromonomers, by living anionic polymerization

Specifications

CAS 2525-62-4
Density 0.873 g/mL (at 25°C (literature))
Boiling Point 162°C to 164°C (lit.)
Molecular Formula C7H13NO
Refractive Index n20/D 1.421 (literature)
Linear Formula CH3(CH2)5 NCO
MDL Number MFCD00037047
Quantity 5 mL
Molecular Weight (g/mol) 127.18
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.