Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hexyl nicotinate | 23597-82-2 | 99.0% | 1 ML
SDP

Catalog No. 5000234823
Encompass_Preferred

Hexyl nicotinate (nicotinic acid hexyl ester) is a nicotinic acid ester used as a topical vasodilator and research reagent. This product is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) for laboratory research, with other solid and liquid pack sizes available.

  • 10 mM solution in DMSO, 1 mL vial.
  • CAS number 23597-82-2.
  • Molecular weight 207.27 g/mol; formula C12H17NO2.
  • Stated purity 98.96%.
  • Intended for research use only.

Catalog No. 50-002-34823 Supplier Medchemexpress LLC Supplier No. HY12166010MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hexyl nicotinate (nicotinic acid hexyl ester) is a nicotinic acid ester used as a topical vasodilator and research reagent. This product is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) for laboratory research, with other solid and liquid pack sizes available.

  • 10 mM solution in DMSO, 1 mL vial.
  • CAS number 23597-82-2.
  • Molecular weight 207.27 g/mol; formula C12H17NO2.
  • Stated purity 98.96%.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.