Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hexyl nicotinate | 23597-82-2 | 99.0% | 100 MG
SDP

Catalog No. 5000234819
Encompass_Preferred

Hexyl nicotinate is a nicotinic acid ester used as a laboratory reagent in biochemical and pharmacological research. It is typically supplied as a liquid with documented purity, molecular properties, and defined storage and solubility recommendations.

  • Purity 98.96%.
  • Molecular formula C12H17NO2.
  • Molecular weight 207.27 g/mol.
  • Appearance colorless to light yellow liquid; density 1.02 g/cm3.
  • Solubility DMSO soluble at 100 mg/mL; in vivo formulation ≥ 2.5 mg/mL in 10% DMSO with SBE-β-CD in saline.
  • Storage pure form: -20°C (3 years) or 4°C (2 years); in solvent: -80°C (6 months).

Catalog No. 50-002-34819 Supplier Medchemexpress LLC Supplier No. HY121660100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hexyl nicotinate is a nicotinic acid ester used as a laboratory reagent in biochemical and pharmacological research. It is typically supplied as a liquid with documented purity, molecular properties, and defined storage and solubility recommendations.

  • Purity 98.96%.
  • Molecular formula C12H17NO2.
  • Molecular weight 207.27 g/mol.
  • Appearance colorless to light yellow liquid; density 1.02 g/cm3.
  • Solubility DMSO soluble at 100 mg/mL; in vivo formulation ≥ 2.5 mg/mL in 10% DMSO with SBE-β-CD in saline.
  • Storage pure form: -20°C (3 years) or 4°C (2 years); in solvent: -80°C (6 months).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.