Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Hexylbenzene

Catalog No. P2570125G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2570125G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P2570125G Supplier Sigma Aldrich Supplier No. P2570125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-(2-aminoethyl)-1-methylpyrrolidine labelling acts as probe within microfluidic chip for electrochemiluminescence detection
  • It acts as electrogenerated chemiluminescence derivatization reagent for carboxylic acid in HPLC.2-(2-aminoethyl)-1-methylpyrrolidine was used in dual-cloud point extraction and tertiary amine labeling for the quantification of indole-3-acetic acid and indole-3-butyric acid

Specifications

CAS 1077-16-3
Melting Point -61°C (lit.)
Density 0.861 g/mL (at 25°C (literature))
Boiling Point 226°C (lit.)
Molecular Formula C12H18
Refractive Index n20/D 1.486 (literature)
Linear Formula C6H5(CH2)5 CH3
MDL Number MFCD00009526
Quantity 1 g
Synonym 1-Phenylhexane
Molecular Weight (g/mol) 162.27
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.