Learn More
Medchemexpress LLC Hexylene glycol | 107-41-5 | 98.0% | 1 ML
SDP
Catalog No. 5000409062
Encompass_Preferred
Hexylene glycol is a low molecular weight surfactant with antimicrobial properties. It is widely used in industrial chemicals and cosmetics.
  • Low molecular weight surfactant
  • Possesses antimicrobial properties
  • Widely used in industrial chemicals
  • Widely used in cosmetic applications
  • Effective against various microorganisms including Candida albicans, Staphylococcus aureus, and Escherichia coli
Catalog No. 50-004-09062 Supplier Medchemexpress LLC Supplier No. HYB090310MM/1ML
Hexylene glycol is a low molecular weight surfactant with antimicrobial properties. It is widely used in industrial chemicals and cosmetics.
  • Low molecular weight surfactant
  • Possesses antimicrobial properties
  • Widely used in industrial chemicals
  • Widely used in cosmetic applications
  • Effective against various microorganisms including Candida albicans, Staphylococcus aureus, and Escherichia coli