Learn More
Medchemexpress LLC Hexylene glycol | 107-41-5 | MFCD00004547 | 98.0% | 1 G
SDP
Catalog No. 5000409196
Encompass_Preferred
Hexylene glycol is a low molecular weight surfactant with antimicrobial properties. It is widely used in industrial chemicals and cosmetics.
  • Low molecular weight surfactant
  • Antimicrobial properties
  • Widely used in industrial chemicals and cosmetics
  • At 10% and 30% concentrations, it killed *Candida albicans*, *Staphylococcus aureus*, *Staphylococcus epidermidis*, Group A Streptococcus, *Streptococcus parvus*, and *Escherichia coli* within 20 hours *in vitro*
  • At 5% concentration, it showed some antimicrobial activity
Catalog No. 50-004-09196 Supplier Medchemexpress LLC Supplier No. HYB09031G
Hexylene glycol is a low molecular weight surfactant with antimicrobial properties. It is widely used in industrial chemicals and cosmetics.
  • Low molecular weight surfactant
  • Antimicrobial properties
  • Widely used in industrial chemicals and cosmetics
  • At 10% and 30% concentrations, it killed *Candida albicans*, *Staphylococcus aureus*, *Staphylococcus epidermidis*, Group A Streptococcus, *Streptococcus parvus*, and *Escherichia coli* within 20 hours *in vitro*
  • At 5% concentration, it showed some antimicrobial activity