Learn More
Selleck Chemical LLC Hexylene glycol S3588-100mg
SDP
Catalog No. 503109345
Encompass_Preferred
Hexylene glycol (HG 2-Methyl-2 4-pentanediol MPD) is an oxygenated solvent derived from acetone that has been used widely in industrial chemicals and cosmetics Hexylene glycol exhibits antibacterial and antifungal properties
Catalog No. 50-310-9345 Supplier Selleck Chemical LLC Supplier No. S3588100MG
Hexylene glycol (HG 2-Methyl-2 4-pentanediol MPD) is an oxygenated solvent derived from acetone that has been used widely in industrial chemicals and cosmetics Hexylene glycol exhibits antibacterial and antifungal properties