Learn More
Cayman Chemical Hexylglutathione, Purity: ≥95%, CAS: 24425-56-7, 100 mg
SDP
Catalog No. 502992573
Encompass_Preferred
An mtMGST1 inhibitor (IC50 = 0.48 mM); inhibits the dopachrome tautomerase activity of MIF (IC50 = 3.3 mM); has been used as an affinity chromatography ligand to purify GST from sheep liver homogenate
Catalog No. 50-299-2573 Supplier Cayman Chemical Supplier No. 29695100 MG
An mtMGST1 inhibitor (IC50 = 0.48 mM); inhibits the dopachrome tautomerase activity of MIF (IC50 = 3.3 mM); has been used as an affinity chromatography ligand to purify GST from sheep liver homogenate