Learn More
Apexbio Technology LLC Hexylresorcinol 136-77-6 1g
SDP
Catalog No. 502240703
Encompass_Preferred
Hexylresorcinol (136-77-6) is a small-molecule inhibitor targeting mushroom tyrosinase a copper-containing enzyme catalyzing the oxidation of phenolic substrates It is designed to modulate tyrosinase activity thereby regulating melanogenesis pathways Hexylresorcinol exerts its biological activity primarily through competitive inhibition of diphenolase activity In in vitro studies Hexylresorcinol demonstrates inhibition of both monophenolase and diphenolase activities with an IC50 value of approximately 1 24 M and a Ki of about 0 443 M Based on these pharmacological properties Hexylresorcinol holds research potential in pigmentation regulation therapeutic strategies for hyperpigmentation disorders and has clinical applications in topical formulations for skin infections throat lozenges and anti-aging skincare products
This item is not returnable. View return policy
Catalog No. 50-224-0703 Supplier Apexbio Technology LLC Supplier No. B61331000
Hexylresorcinol (136-77-6) is a small-molecule inhibitor targeting mushroom tyrosinase a copper-containing enzyme catalyzing the oxidation of phenolic substrates It is designed to modulate tyrosinase activity thereby regulating melanogenesis pathways Hexylresorcinol exerts its biological activity primarily through competitive inhibition of diphenolase activity In in vitro studies Hexylresorcinol demonstrates inhibition of both monophenolase and diphenolase activities with an IC50 value of approximately 1 24 M and a Ki of about 0 443 M Based on these pharmacological properties Hexylresorcinol holds research potential in pigmentation regulation therapeutic strategies for hyperpigmentation disorders and has clinical applications in topical formulations for skin infections throat lozenges and anti-aging skincare products