Learn More
LGC GENOMICS LLC Hexynyl CE-Phosphoramidite, Versatile Reagent for Click Chemistry, 1 g
Catalog No. NC3449222
Encompass
Hexynyl CE-Phosphoramidite is a versatile reagent designed for Click Chemistry, facilitating the introduction of hexynyl groups into oligonucleotides. This unique feature allows for efficient conjugation and labeling, enhancing molecular biology and diagnostic applications. Its 1 g weight and MerMade bottle type ensure easy handling and precise usage in various workflows.
Catalog No. NC3449222 Supplier LGC GENOMICS LLC Supplier No. BA0284C0011G
Hexynyl CE-Phosphoramidite is a versatile reagent designed for Click Chemistry, facilitating the introduction of hexynyl groups into oligonucleotides. This unique feature allows for efficient conjugation and labeling, enhancing molecular biology and diagnostic applications. Its 1 g weight and MerMade bottle type ensure easy handling and precise usage in various workflows.