Learn More
Cayman Chemical Hexythiazox, Purity: ≥98%, CAS: 78587-05-0, 100 mg
SDP
Catalog No. 502935909
Encompass_Preferred
An acaricide; induces toxicity in larvae of the two-spotted spider mite T. urticae and the European red mite P. ulmi (LC50s = 0.15-0.58 and 0.23-0.62 mg AI/L, respectively), as well as in the summer and winter eggs of P. ulmi (LC50s = 2.2 and 20 ppm, respectively); toxic to bluegill (L. macrochirus; LC50 = 3.2 mg/L) and D. magna (EC50 = 0.36 mg/L) but not rats (LD50 = >5,000 mg/kg)
Catalog No. 50-293-5909 Supplier Cayman Chemical Supplier No. 25626100 MG
An acaricide; induces toxicity in larvae of the two-spotted spider mite T. urticae and the European red mite P. ulmi (LC50s = 0.15-0.58 and 0.23-0.62 mg AI/L, respectively), as well as in the summer and winter eggs of P. ulmi (LC50s = 2.2 and 20 ppm, respectively); toxic to bluegill (L. macrochirus; LC50 = 3.2 mg/L) and D. magna (EC50 = 0.36 mg/L) but not rats (LD50 = >5,000 mg/kg)