Learn More
Apexbio Technology LLC HEXYTHIAZOX-250MG
SDP
Catalog No. 5000570269
Encompass_Preferred
Hexythiazox (CAS No 78587-05-0) is a chemical compound used primarily as an acaricide in biomedical research targeting mite growth and development Originating from the thiazolidinone class it acts by inhibiting the mitochondrial electron transport chain effectively targeting the mite s energy metabolism Its mechanism of action involves disrupting chitin synthesis crucial for mite exoskeleton formation Common in vitro activity ranges from nanomolar to low micromolar concentrations reflecting its potency Hexythiazox is used in various applications including cell models and animal studies to determine its efficacy and safety It is valuable in drug screening processes to explore potential acaricidal properties against resistant mite strains Experimental concentrations typically depend on the specific research design allowing flexibility in application based on targeted outcomes
Catalog No. 50-005-70269 Supplier Apexbio Technology LLC Supplier No. BA2166250
Hexythiazox (CAS No 78587-05-0) is a chemical compound used primarily as an acaricide in biomedical research targeting mite growth and development Originating from the thiazolidinone class it acts by inhibiting the mitochondrial electron transport chain effectively targeting the mite s energy metabolism Its mechanism of action involves disrupting chitin synthesis crucial for mite exoskeleton formation Common in vitro activity ranges from nanomolar to low micromolar concentrations reflecting its potency Hexythiazox is used in various applications including cell models and animal studies to determine its efficacy and safety It is valuable in drug screening processes to explore potential acaricidal properties against resistant mite strains Experimental concentrations typically depend on the specific research design allowing flexibility in application based on targeted outcomes