Learn More
Medchemexpress LLC Hexythiazox | 78587-05-0 | 99.6% | 250 MG
SDP
Catalog No. 5000414336
Encompass_Preferred
Hexythiazox is a selective acaricide with ovicidal, larvicidal, and nymphicidal activities. It is widely used for the chemical control of mites on cotton, fruits, and vegetables. This product is harmless to mammals and does not affect beneficial insects and predators of mites.
  • Selective acaricide.
  • Exhibits ovicidal activity.
  • Exhibits larvicidal activity.
  • Exhibits nymphicidal activity.
  • Widely used for mite control on cotton, fruits, and vegetables.
  • Harmless to mammals.
  • No effect on beneficial insects and predators of mites.
Catalog No. 50-004-14336 Supplier Medchemexpress LLC Supplier No. 250MGHYB1851
Hexythiazox is a selective acaricide with ovicidal, larvicidal, and nymphicidal activities. It is widely used for the chemical control of mites on cotton, fruits, and vegetables. This product is harmless to mammals and does not affect beneficial insects and predators of mites.
  • Selective acaricide.
  • Exhibits ovicidal activity.
  • Exhibits larvicidal activity.
  • Exhibits nymphicidal activity.
  • Widely used for mite control on cotton, fruits, and vegetables.
  • Harmless to mammals.
  • No effect on beneficial insects and predators of mites.