Learn More
Aviva Systems Biology Corp HEY1 Antibody - C-terminal region (ARP32512_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32512_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against HEY1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human HEY1. Alternate names: CHF2, OAF1, HERP2, HESR1, HRT-1, BHLHb31ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54628