Learn More
Aviva Systems Biology Corp HEY1 Antibody - middle region (ARP41453_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41453_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HEY1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HEY1. Alternate names: CHF2, HERP2, HESR1, HRT-1, MGC1274, OAF1, BHLHb31ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57864