Learn More
Aviva Systems Biology Corp HEY2 Antibody - middle region (ARP38979_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38979_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HEY2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HEY2. Alternate names: CHF1, GRL, HERP1, HESR2, HRT2, MGC10720, bHLHb32, GRIDLOCKApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56878