Learn More
Aviva Systems Biology Corp HEYL Antibody - N-terminal region (ARP31866_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31866_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against HEYL. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HEYL. Alternate names: HEY3, HRT3, bHLHb33ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-54348