Learn More
Cole-Parmer Cole-Parmer Hammer-forged aluminum Dewar flask, 14 L
Supplier:  Cole-Parmer 0377456
Encompass_Preferred
Durable construction and easy-to-grip handle is perfect for field and bench use. Use for short-term storage or safely transport liquid nitrogen, dry ice, or cryogens. Hammer forged aluminum casing exterior. The cushioned borosilicate glass inner vessel resists breakage. Insulating lid and fold-down handle.
Catalog No. NC1290476