Learn More
Glass Expansion HF Mixing T-Piece
SDP
Catalog No. 502703091
Encompass_Preferred
HF Mixing T-Piece. Replacement HF Mixing T-Piece for Glass Expansion’s Trident IS Kit for HF P/N NC2046964, 60-808-1150.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3091 Supplier Glass Expansion Supplier No. 608081124
HF Mixing T-Piece. Replacement HF Mixing T-Piece for Glass Expansion’s Trident IS Kit for HF P/N NC2046964, 60-808-1150.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001