Learn More
HF Scientific™ Chlorine Micro Check Test Strips
Catalog No. 14376164
Click to view available options
Type:
Free ChlorineTest Strip
Catalog No. Type
14-376-164 Free ChlorineTest Strip
1 options
Catalog No. 14-376-164 Supplier HF Scientific™ Supplier No. 09940
Only null left
Add to Cart
Add to Cart
Includes
50 test strips are supplied in bottle labeled with instructions, color-comparison scale, and expiration date
Detect free or total chlorine concentrations as low as 0.05ppm
Test strips for field testing and quality-control procedures; provide a safe and economical alternative to reagent chlorine testing.
  • Accurate results produced in less than one minute
  • Easy to use—simply dip strip into sample or hold under liquid stream to saturate, remove, and match color to the color-comparison scale
Specifications
Format Strip
For Use With (Application) For detection of total chlorine, field testing and quality-control procedures, provide a safe and economical alternative to reagent chlorine testing
Includes 50 test strips are supplied in bottle labeled with instructions, color comparison scale, and expiration date
Quantity 50 Pack
Range 0, 0.1, 0.2, 0.4, 0.6, 0.8, 1.2, 1.5, 2.0, 2.6, 4.0, 6.0, 10
Test Time >1 min.
Type Free ChlorineTest Strip