Learn More
HF Scientific™ Formazin Stock Solution Kit
Catalog No. 153932
Click to view available options
Type:
Formazin Stock Solution Kit
Catalog No. Type
15-393-2 Formazin Stock Solution Kit
1 options
Catalog No. 15-393-2 Supplier HF Scientific™ Supplier No. 50040
Can be used with DRT-15CE Portable Turbidimeter and Micro 100 Laboratory Turbidimeter. Contains 1 gal. (3.8L) water (with a 0.02 NTU value), 32 oz. (946mL) stock (4000 NTU), one 0.02 NTU reference standard, four cuvettes with light shield caps, three pipets, and a 20-page instruction manual for making standard values for turbidimeter calibration.
Specifications
Type Formazin Stock Solution Kit
For Use With (Equipment) DRT-15CE Portable Turbidimeter and Micro 100 Laboratory Turbidimeter
Includes 946mL of 4000 NTU formazin, 3.8L 0.02 NTU reference standard, 3.8L of 0.02 NTU water, 4 cuvettes with indexing light shields, 3 pipettes and instruction manual