Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HF Scientific™ Powder Pop™ DPD Reagent for Chlorine Testing

Catalog No. p-4363636
Encompass
Change view
Click to view available options
Quantity:
1000/Pk.
200/Pk.
Volume (Metric):
5 mL
10 mL
25 mL
For Use With (Application):
For testing free chlorine
For testing total chlorine
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Volume (Metric) For Use With (Application)
23-285188 200/Pk. 10 mL For testing total chlorine
23-285184 200/Pk. 5 mL For testing total chlorine
15-393-67 200/Pk. 25 mL For testing total chlorine
15-393-58 1000/Pk. 5 mL For testing free chlorine
15-393-60 1000/Pk. 5 mL For testing total chlorine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 23-285188 Supplier HF Scientific™ Supplier No. 10309
Only null left
Add to Cart
Add to Cart

For testing free or total chlorine

  • Deliver doses of DPD reagent measured for 5, 10 or 25mL (0.2, 0.3 or 0.8 oz.) testing samples
  • Individually packaged

Specifications

Volume (Metric) 10 mL
For Use With (Application) For testing total chlorine
Quantity 200/Pk.
Type DPD Reagent
Volume (English) 0.33 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.