Learn More
HF Scientific™ Procal Primary Calibration Kit
Catalog No. 1539356
Click to view available options
Turbidity:
0.02, 10, 1000 NTU
Catalog No. Turbidity
15-393-56 0.02, 10, 1000 NTU
1 options
Catalog No. 15-393-56 Supplier HF Scientific™ Supplier No. 39845
Specifications
Type Primary Calibration Kit
For Use With (Equipment) MicroTPI and MicroTPW Portable Turbidimeters
Turbidity 0.02, 10, 1000 NTU