Learn More
Teledyne ISCO, Inc Hf Series Pump, 300Ml; 10,000 Psi; Stainless Steel, Self-Flush, With Pulse Dampener And Transducer, Constant Flow, Large Motor
Catalog No. 502119937 Shop All Teledyne ISCO Inc. Products
Encompass
HF Series Pump, 300mL; 10,000 psi; Stainless Steel, Self-Flush, with Pulse Dampener and Transducer, Constant Flow, Large Motor
Catalog No. 50-211-9937 Supplier Teledyne ISCO, Inc Supplier No. HF300SRX
Add to Cart
Add to Cart
Request a Quote
HF Series Pump, 300mL; 10,000 psi; Stainless Steel, Self-Flush, with Pulse Dampener and Transducer, Constant Flow, Large Motor