Learn More
Calgonate HF SPILLKIT- SINGLE
Catalog No. NC0581502
Encompass_Preferred
The 5-gallon bucket contains all the personal protection equipment and supplies for 1 person to safely deal with an HF acid spill. Including 1 Tube of Calgonate Gel and 1 Bottle of Calgonate Emergency Eyewash and 4lbs of HF neutralizer
Catalog No. NC0581502 Supplier Calgonate Supplier No. KITSPILL01
The 5-gallon bucket contains all the personal protection equipment and supplies for 1 person to safely deal with an HF acid spill. Including 1 Tube of Calgonate Gel and 1 Bottle of Calgonate Emergency Eyewash and 4lbs of HF neutralizer