Learn More
Calgonate HF SPILLKIT
Catalog No. NC0279527
Encompass_Preferred
The 2 5-gallon buckets contain all the personal protection equipment and supplies for 2 people to safely deal with a HF acid spill. Including 2 Tubes of Calgonate Gel and 2 Bottles of Calgonate Emergency Eyewash and 9lbs of HF neutralizer
Catalog No. NC0279527 Supplier Calgonate Supplier No. KITSPILL02
The 2 5-gallon buckets contain all the personal protection equipment and supplies for 2 people to safely deal with a HF acid spill. Including 2 Tubes of Calgonate Gel and 2 Bottles of Calgonate Emergency Eyewash and 9lbs of HF neutralizer