Learn More
As One International Inc High Fidelity Taq Polymerase, 10x Ammonium Buffer, Tween free, 1,000U
Catalog No. 501655861
Encompass_Preferred
AS ONE PR Taq DNA Polymerase is a thermostable high fidelity DNA polymerase with proof reading ability. This feature enables accurate and reliable PCR. Besides a 5' to 3' polymerase activity, PR-Taq exhibits a 3' to 5' exonuclease activity that enables the enzyme to correct base pair mismatches. This results in PCR products with very few errors and blunt ends. PR Taq Polymerase includes Tween free 10x Ammonium Buffer.
Catalog No. 50-165-5861 Supplier As One International Inc Supplier No. AO211404
AS ONE PR Taq DNA Polymerase is a thermostable high fidelity DNA polymerase with proof reading ability. This feature enables accurate and reliable PCR. Besides a 5' to 3' polymerase activity, PR-Taq exhibits a 3' to 5' exonuclease activity that enables the enzyme to correct base pair mismatches. This results in PCR products with very few errors and blunt ends. PR Taq Polymerase includes Tween free 10x Ammonium Buffer.