Learn More
Aviva Systems Biology Corp HFE Antibody - N-terminal region (ARP44283_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44283_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HFE. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HFE. Alternate names: HFE1, HH, HLA-H, MGC103790, dJ221C16.10.1, MVCD7, TFQTL2ApplicationWB. Species reactivity: Cow, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-133