Learn More
ATCC HFF-1 fibroblast; IRR; Human foreskin
Catalog No. 502384469
Encompass
HFF-1 IRR is a fibroblast cell that was isolated from the foreskin of a donor. The irradiated cells can be used as feeder cells to support the growth of stem cells in the undifferentiated state.
Catalog No. 50-238-4469 Supplier ATCC Supplier No. SCRC1041.1
HFF-1 IRR is a fibroblast cell that was isolated from the foreskin of a donor. The irradiated cells can be used as feeder cells to support the growth of stem cells in the undifferentiated state.