Learn More
Lonza Walkersville hFGF – Human Fibroblastic Growth Factor, 1 mL
Supplier:  Lonza Walkersville CC4068
Encompass
Fibroblast growth factors (FGFs), is a combination of growth factors involved in angiogenesis, wound healing, embryonic development and various endocrine signaling pathways. The FGFs are heparin-binding proteins and interactions with cell-surface-associated heparan sulfate proteoglycans have been shown to be essential for FGF signal transduction. FGFs are key players in the processes of proliferation and differentiation of wide variety of cells and tissues. The Fibroblastic Growth Factor, recombinant is available at a concentration of 1µg/ml.Content:1 x Vial Containing 1 ug/mL hFGF, 1 mL
Catalog No. NC2389439