Learn More
Regis Technologies Inc GC Regeant, Regis, HFIP (100 GM)
SDP
Supplier:  Regis Technologies Inc 1270704200
Encompass
HFIP is an Acylation Reagent. Esterification reagent for the determination of aromatic acids in tissue by GC and electron capture detection (ECD).
Catalog No. 50-442-928