Learn More
Medchemexpress LLC HFY-4A | 2094810-82-7 | 99.3% | 333.38 | 25 MG
SDP
Catalog No. 5000361259
Encompass_Preferred
HFY-4A is an HDAC inhibitor that inhibits breast cancer cell proliferation, migration, and invasion, and induces cell apoptosis. It also induces immunogenic cell death (ICD) and inhibits tumor growth in breast cancer xenograft mouse models.
  • Inhibits breast cancer cell proliferation, migration, and invasion
  • Induces cell apoptosis
  • Induces immunogenic cell death (ICD)
  • Inhibits tumor growth in breast cancer xenograft mouse models
Catalog No. 50-003-61259 Supplier Medchemexpress LLC Supplier No. 25MGHY149631
HFY-4A is an HDAC inhibitor that inhibits breast cancer cell proliferation, migration, and invasion, and induces cell apoptosis. It also induces immunogenic cell death (ICD) and inhibits tumor growth in breast cancer xenograft mouse models.
  • Inhibits breast cancer cell proliferation, migration, and invasion
  • Induces cell apoptosis
  • Induces immunogenic cell death (ICD)
  • Inhibits tumor growth in breast cancer xenograft mouse models