Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HFY-4A | 2094810-82-7 | 99.3% | 333.38 | 50 MG
SDP

Catalog No. 5000360807
Encompass_Preferred

HFY-4A is a Histone Deacetylase (HDAC) inhibitor. It inhibits breast cancer cell proliferation, migration, and invasion, and induces cell apoptosis and immunogenic cell death (ICD). It also inhibits tumor growth in breast cancer xenograft mouse models.

  • HDAC inhibitor
  • Inhibits breast cancer cell proliferation, migration, and invasion
  • Induces cell apoptosis and immunogenic cell death (ICD)
  • Inhibits tumor growth in breast cancer xenograft mouse models
  • Purity: 99.34%
  • Form: Solid
  • Color: White to off-white
  • Soluble in DMSO

Catalog No. 50-003-60807 Supplier Medchemexpress LLC Supplier No. HY14963150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

HFY-4A is a Histone Deacetylase (HDAC) inhibitor. It inhibits breast cancer cell proliferation, migration, and invasion, and induces cell apoptosis and immunogenic cell death (ICD). It also inhibits tumor growth in breast cancer xenograft mouse models.

  • HDAC inhibitor
  • Inhibits breast cancer cell proliferation, migration, and invasion
  • Induces cell apoptosis and immunogenic cell death (ICD)
  • Inhibits tumor growth in breast cancer xenograft mouse models
  • Purity: 99.34%
  • Form: Solid
  • Color: White to off-white
  • Soluble in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.