Learn More
Cayman Chemical HG-10-102-01 1mg
SDP
Catalog No. 502984378
Encompass_Preferred
An inhibitor of LRRK2 (IC50 = 20.3 nM); also inhibits G2019S, A2016T, and [G2019S+A2016T] mutants of LRRK2 (IC50s = 3.2, 153, and 95.9 nM, respectively); brain penetrant and inhibits Ser910 and Ser935 phosphorylation of both wild-type and G2019S mutant LRRK2
Catalog No. 50-298-4378 Supplier Cayman Chemical Supplier No. 200491 MG
An inhibitor of LRRK2 (IC50 = 20.3 nM); also inhibits G2019S, A2016T, and [G2019S+A2016T] mutants of LRRK2 (IC50s = 3.2, 153, and 95.9 nM, respectively); brain penetrant and inhibits Ser910 and Ser935 phosphorylation of both wild-type and G2019S mutant LRRK2