Learn More
Medchemexpress LLC HY-15801 5mg Medchemexpress, HG-14-10-04 CAS:1356962-34-9 Purity:>98%
SDP
Catalog No. 502081801
Encompass_Preferred
Medchemexpress, HY-15801 5mg HG-14-10-04 CAS:1356962-34-9 HG-14-10-04 (example 10) is a potent and specific ALK inhibitor with an IC50 of 20 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1801 Supplier Medchemexpress LLC Supplier No. HY158015MG
Medchemexpress, HY-15801 5mg HG-14-10-04 CAS:1356962-34-9 HG-14-10-04 (example 10) is a potent and specific ALK inhibitor with an IC50 of 20 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.