Learn More
Medchemexpress LLC HG-7-85-01-NH2 | 1258391-29-5 | 98.1% | 50 MG
SDP
Catalog No. 5000338550
Encompass_Preferred
HG-7-85-01-NH2 is a small-molecule ligand derived from an ABL inhibitor. It serves as a target protein ligand for synthesizing the heterobifunctional molecule SNIPER and can bind to an IAP ligand via a linker to form SNIPER(ABL)-033. The resulting SNIPER(ABL)-033 effectively degrades BCR-ABL protein.
  • Derived from an ABL inhibitor.
  • Functions as a target protein ligand for SNIPER synthesis.
  • Forms a heterobifunctional molecule when linked to an IAP ligand.
  • Used in the formation of SNIPER(ABL)-033, which degrades BCR-ABL protein.
  • Intended for research use only.
Catalog No. 50-003-38550 Supplier Medchemexpress LLC Supplier No. HY13117850MG
HG-7-85-01-NH2 is a small-molecule ligand derived from an ABL inhibitor. It serves as a target protein ligand for synthesizing the heterobifunctional molecule SNIPER and can bind to an IAP ligand via a linker to form SNIPER(ABL)-033. The resulting SNIPER(ABL)-033 effectively degrades BCR-ABL protein.
  • Derived from an ABL inhibitor.
  • Functions as a target protein ligand for SNIPER synthesis.
  • Forms a heterobifunctional molecule when linked to an IAP ligand.
  • Used in the formation of SNIPER(ABL)-033, which degrades BCR-ABL protein.
  • Intended for research use only.