Learn More
Medchemexpress LLC Hg-7-85-01-nh2 | 1258391-29-5 | 98.1% | 540.60 | C27H27F3N6OS | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1311785MG
Encompass_Preferred
HG-7-85-01-NH2 is a small-molecule ligand derived from an ABL inhibitor, designed for use as the target-protein binding moiety in heterobifunctional degraders such as SNIPERs and PROTACs. Supplied as a high-purity solid, the compound includes clear solubility and storage recommendations appropriate for research applications; not for human use.
  • High purity solid suitable for research.
  • Soluble in DMSO at high concentration (125 mg/mL) with pH adjustment guidance.
  • Provides a binding moiety for Bcr-Abl-targeting degraders.
  • Specified storage conditions for powder and in solvent to maintain stability.
  • Intended for research use only.
Catalog No. 50-002-62264