Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HG-7-85-01-NH2 | 1258391-29-5 | MFCD34646287 | 98.1% | C27H27F3N6OS | 10MG
SDP

Catalog No. 5000260475
Encompass_Preferred

HG-7-85-01-NH2 is a research ligand derived from the ABL inhibitor HG-7-85-01, designed to act as the target-protein-binding moiety in SNIPER constructs for targeted protein degradation studies. It is supplied as a characterized, high-purity compound with manufacturer-recommended solubility and storage conditions for preclinical research use.

  • High purity (98.1%).
  • Molecular formula C27H27F3N6OS; molecular weight 540.6.
  • Soluble in DMSO at 125 mg/mL (231.22 mM) with ultrasonic aid and pH adjustment.
  • Powder stable at -20°C for up to 3 years; in solvent stable at -80°C for 6 months.
  • Supplied in multiple small research quantities.
  • Intended for synthesis of SNIPER molecules and targeted degradation assays.

Catalog No. 50-002-60475 Supplier Medchemexpress LLC Supplier No. HY13117810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HG-7-85-01-NH2 is a research ligand derived from the ABL inhibitor HG-7-85-01, designed to act as the target-protein-binding moiety in SNIPER constructs for targeted protein degradation studies. It is supplied as a characterized, high-purity compound with manufacturer-recommended solubility and storage conditions for preclinical research use.

  • High purity (98.1%).
  • Molecular formula C27H27F3N6OS; molecular weight 540.6.
  • Soluble in DMSO at 125 mg/mL (231.22 mM) with ultrasonic aid and pH adjustment.
  • Powder stable at -20°C for up to 3 years; in solvent stable at -80°C for 6 months.
  • Supplied in multiple small research quantities.
  • Intended for synthesis of SNIPER molecules and targeted degradation assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.