Learn More
Medchemexpress LLC HG-9-91-01 | 1456858-58-4 | MFCD28167714 | 10 mM * 1 mL in DMSO
SDP
Supplier:  Medchemexpress LLC HY1577610MM/1ML
Encompass_Preferred
HG-9-91-01 | Putity: 98.17% | MW: 567.68 | 1456858-58-4 | MFCD28167714 | 10 mM * 1 mL in DMSO
Catalog No. NC1368190