Learn More
Apexbio Technology LLC HG-9-91-01, 10mg CAS# 1456858-58-4
SDP
Supplier:  Apexbio Technology LLC B105210
Encompass_Preferred
HG-9-91-01 is a pan-SIK (salt-inducible kinases) inhibitor with IC50 values of 0.92nM, 6.6nM and 9.6nM for SIK1, SIK2, SIK3, respectively [1].SIKs restrict the formation of regulatory macrophages. The inhibition of these enzymes is thought to have potential for treating inflammatory and autoimmune diseases. HG-9-91-01 potently inhibits SIKs through targeting both the ATP-binding site and the small hydrophobic pocket. It is reported that, besides SIKs, HG-9-91-01 also inhibits various protein tyrosine kinases such as Src, BTK, FGF and Ephrin receptors. But other AMPK-related kinase subfamily members are not sensitive to HG-9-91-01. HG-9-91-01 can promote LPS-stimulated IL-10 production, CREB-dependent gene transcription, inhibit proinflammatory cytokine secretion and induce the expression of regulatory macrophage markers. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8384