Learn More
Selleck Chemical LLC HG-9-91-01 5mg
SDP
Supplier:  Selleck Chemical LLC S83935MG
Encompass_Preferred
HG-9-91-01 (SIK inhibitor 1) is a potent and highly selective inhibitor of salt-inducible kinase (SIK) with IC50 of 0.92 nM, 6.6 nM and 9.6 nM for SIK1, SIK2 and SIK3, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3358