Learn More
Sigma Aldrich Fine Chemicals Biosciences Dulbecco's Modified Eagle's Medium - high glucose With 4500 mg/L glucose, L-glutamine, sodium pyruvate, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture | 6X1L
Catalog No. NC1975799
Encompass
Dulbecco's Modified Eagle's Medium - high glucose With 4500 mg/L glucose, L-glutamine, sodium pyruvate, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture | 6X1L
Catalog No. NC1975799 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D64296X1L
Dulbecco's Modified Eagle's Medium - high glucose With 4500 mg/L glucose, L-glutamine, sodium pyruvate, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture | 6X1L