Learn More
Eurofins Discoverx PrecisION hGABAA α5/β3/γ2 Recombinant Stable Cell Line, 2 vials. (CYL3073)
Catalog No. 502005101
Encompass
The PrecisION hGABAA α5/β3/γ2 Recombinant Stable Cell Line is a stable, engineered cell line for use in studying GABAA α5/β3/γ2 ion channels, including drug candidates which affect channel function. This cell line enables assessment of ion channel function via detection of electrophysiological signals. The included cell line overexpresses GABAA α5/β3/γ2 ion channels. Upon ligand/agonist induced channel activation, response can be detected via automated or manual electrophysiological techniques. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/ion-channels.
Catalog No. 50-200-5101 Supplier Eurofins Discoverx Supplier No. CYL3073
Add to Cart
Add to Cart
Request a Quote
The PrecisION hGABAA α5/β3/γ2 Recombinant Stable Cell Line is a stable, engineered cell line for use in studying GABAA α5/β3/γ2 ion channels, including drug candidates which affect channel function. This cell line enables assessment of ion channel function via detection of electrophysiological signals. The included cell line overexpresses GABAA α5/β3/γ2 ion channels. Upon ligand/agonist induced channel activation, response can be detected via automated or manual electrophysiological techniques. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/ion-channels.