Learn More
New England Biolabs, Inc. HgaI – 100 units
SDP
Catalog No. 50812049
Encompass
A restriction endonuclease that recognizes the sequence GACGCNNNNN^NNNNN_.
Catalog No. 50-812-049 Supplier New England Biolabs, Inc. Supplier No. R0154S
A restriction endonuclease that recognizes the sequence GACGCNNNNN^NNNNN_.