Learn More
Eurofins Discoverx HGF, 10 µg. (92-1140)
Catalog No. 501999672
Encompass
Hepatocyte growth factor (HGF) is paracrine growth factor that regulates cell growth, motility, and morphogenesis by activating a tyrosine kinase signaling cascade upon binding to the proto-oncogenic c-Met receptor. Secreted by mesenchymal cells, it has a central role in angiogenesis, tumorogenesis, and tissue regeneration. Acts as an agonist for c-Met. Provided as 10 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9672 Supplier Eurofins Discoverx Supplier No. 921140
Hepatocyte growth factor (HGF) is paracrine growth factor that regulates cell growth, motility, and morphogenesis by activating a tyrosine kinase signaling cascade upon binding to the proto-oncogenic c-Met receptor. Secreted by mesenchymal cells, it has a central role in angiogenesis, tumorogenesis, and tissue regeneration. Acts as an agonist for c-Met. Provided as 10 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.