Learn More
Aviva Systems Biology Corp HGF Antibody - middle region (ARP44319_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44319_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HGF. It was validated on Western Blot. HGF,hepatocyte growth factor (hepapoietin A; scatter factor),SF; HGFB; HPTA; F-TCF; DFNB39; HGF,hepatocyte growth factor,hepatocyte growth factor,hepatopoeitin-A,OTTHUMP00000206710,OTTHUMP00000206730,lung fibroblast-derived mitogen,fibroblast-derived tumor cytotoxic factorThe immunogen is a synthetic peptide directed towards the middle region of human HGF. Alternate names: F-TCF, HGFB, HPTA, SF, DFNB39ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-137