Learn More
Aviva Systems Biology Corp HGF Antibody - N-terminal region (ARP44317_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44317_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HGF. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HGF. Alternate names: F-TCF, HGFB, HPTA, SF, DFNB39ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-58778